ENA018105797 Enamine

3-amino-N-[(furan-2-yl)methyl]benzene-1-sulfonamide

  • MDL number MFCD02699973
  • Cas Number : 326610-71-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 50-51
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02115

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02115
Availability details for
Quantity
  • Availability
  • Add to Cart