ENA018105800 Enamine

3-Amino-N-(4-methoxy-phenyl)-benzenesulfonamide

  • MDL number MFCD00188538
  • Cas Number : 201996-24-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 109...111
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02118

Safety Information

msds https://www.enaminestore.com/catalog/EN300-02118
Availability details for
Quantity
  • Availability
  • Add to Cart