ENA018105803 Enamine

3-amino-N-(2-chlorophenyl)benzene-1-sulfonamide

  • MDL number MFCD02708234
  • Cas Number : 327092-99-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 110-111
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02121

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02121
Availability details for
Quantity
  • Availability
  • Add to Cart