ENA018105806 Enamine

5-Amino-N-(3-chloro-phenyl)-2-methyl-benzenesulfonamide

  • MDL number MFCD02703353
  • Cas Number : 326871-18-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 158...160
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02124

Safety Information

msds https://www.enaminestore.com/catalog/EN300-02124
Availability details for
Quantity
  • Availability
  • Add to Cart