ENA018105810 Enamine

3-Amino-4-chloro-N,N-dimethyl-benzenesulfonamide

  • MDL number MFCD02704404
  • Cas Number : 100313-81-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 119...121
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02128

Safety Information

msds https://www.enaminestore.com/catalog/EN300-02128
Availability details for
Quantity
  • Availability
  • Add to Cart