ENA018105841 Enamine

3-amino-N-(4-chlorophenyl)-4-methylbenzene-1-sulfonamide

  • MDL number MFCD02704644
  • Cas Number : 328028-61-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02159

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02159
Availability details for
Quantity
  • Availability
  • Add to Cart