ENA018105848 Enamine

2-(morpholin-4-yl)-5-(morpholine-4-sulfonyl)aniline

  • MDL number MFCD02697854
  • Cas Number : 325748-56-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 210-211
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02166

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02166
Availability details for
Quantity
  • Availability
  • Add to Cart