ENA018105851 Enamine

3-amino-N-(4-methoxyphenyl)-4-(piperidin-1-yl)benzene-1-sulfonamide

  • MDL number MFCD02701333
  • Cas Number : 326092-19-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 111-112
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02169

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02169
Availability details for
Quantity
  • Availability
  • Add to Cart