ENA018105856 Enamine

3-Amino-N-(2-chloro-phenyl)-4-morpholin-4-yl-benzenesulfonamide

  • MDL number MFCD02700609
  • Cas Number : 326023-05-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 43...45
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02174

Safety Information

msds https://www.enaminestore.com/catalog/EN300-02174
Availability details for
Quantity
  • Availability
  • Add to Cart