ENA018105863 Enamine

5-amino-N-(2-methoxyphenyl)-2-(piperidin-1-yl)benzene-1-sulfonamide

  • MDL number MFCD02700602
  • Cas Number : 326022-99-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 145-146
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-02181

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02181
Availability details for
Quantity
  • Availability
  • Add to Cart