ENA018105866 Enamine

5-amino-N-(2-methoxyphenyl)-2-(morpholin-4-yl)benzene-1-sulfonamide

  • MDL number MFCD02700620
  • Cas Number : 326023-21-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 160-161
Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02184

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02184
Availability details for
Quantity
  • Availability
  • Add to Cart