ENA018105867 Enamine

5-amino-N-(2-chlorophenyl)-2-(piperidin-1-yl)benzene-1-sulfonamide

  • MDL number MFCD02706748
  • Cas Number : 327072-89-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 135-136
Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02185

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02185
Availability details for
Quantity
  • Availability
  • Add to Cart