ENA018105868 Enamine

5-amino-N-(2-chlorophenyl)-2-(morpholin-4-yl)benzene-1-sulfonamide

  • MDL number MFCD02704646
  • Cas Number : 328028-63-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 170-171
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02186

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02186
Availability details for
Quantity
  • Availability
  • Add to Cart