ENA018105869 Enamine

5-amino-2-(pyrrolidin-1-yl)-N-[3-(trifluoromethyl)phenyl]benzene-1-sulfonamide

  • MDL number MFCD02701688
  • Cas Number : 326182-65-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 125-126
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02187

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02187
Availability details for
Quantity
  • Availability
  • Add to Cart