ENA018105873 Enamine

3-amino-4-hydroxy-N-(2-methoxyphenyl)benzene-1-sulfonamide

  • MDL number MFCD02704606
  • Cas Number : 109568-85-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-02191

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02191
Availability details for
Quantity
  • Availability
  • Add to Cart