ENA018105876 Enamine

3-amino-N-(3-chlorophenyl)-4-hydroxybenzene-1-sulfonamide

  • MDL number MFCD02704631
  • Cas Number : 328028-38-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02194

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02194
Availability details for
Quantity
  • Availability
  • Add to Cart