ENA018105877 Enamine

3-amino-N-(2-chlorophenyl)-4-hydroxybenzene-1-sulfonamide

  • MDL number MFCD02704645
  • Cas Number : 104095-22-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02195

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02195
Availability details for
Quantity
  • Availability
  • Add to Cart