ENA018105894 Enamine

1-Benzyl-2-imino-10-oxo-1,10-dihydro-2H-1,9,10a-triaza-anthracene-3-carbonitrile

  • MDL number MFCD02673362
  • Cas Number : 496799-83-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02214

Safety Information

msds https://www.enaminestore.com/catalog/EN300-02214
Availability details for
Quantity
  • Availability
  • Add to Cart