ENA018105907 Enamine

4-(3-aminophenyl)-1,3-thiazol-2-amine

  • MDL number MFCD03015584
  • Cas Number : 103740-34-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 160-161
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-02230

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02230
Availability details for
Quantity
  • Availability
  • Add to Cart