ENA018105924 Enamine

4-(2,5-dichlorothiophen-3-yl)-1,3-thiazol-2-amine

  • MDL number MFCD00105088
  • Cas Number : 325988-82-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02250

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02250
Availability details for
Quantity
  • Availability
  • Add to Cart