ENA018105947 Enamine

N-(2-amino-1,3-benzothiazol-6-yl)acetamide

  • MDL number MFCD00465253
  • Cas Number : 22307-44-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02275

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02275
Availability details for
Quantity
  • Availability
  • Add to Cart