ENA018105990 Enamine

4-Amino-N-(2-methoxy-phenyl)-benzamide

  • MDL number MFCD00568219
  • Cas Number : 787-59-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 162...164
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02322

Safety Information

msds https://www.enaminestore.com/catalog/EN300-02322
Availability details for
Quantity
  • Availability
  • Add to Cart