ENA018105991 Enamine

4-amino-N-(4-methoxyphenyl)benzamide

  • MDL number MFCD00578773
  • Cas Number : 891-35-0
  • Molecular Weight : 242.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02323

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02323
Availability details for
Quantity
  • Availability
  • Add to Cart