ENA018105998 Enamine

4-amino-N-[3-(trifluoromethyl)phenyl]benzene-1-sulfonamide

  • MDL number MFCD00204069
  • Cas Number : 339-40-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-02330

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02330
Availability details for
Quantity
  • Availability
  • Add to Cart