ENA018106002 Enamine

ethyl 3-amino-4-(ethylamino)benzoate

  • MDL number MFCD00187201
  • Cas Number : 202131-30-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 100-107
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02334

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02334
Availability details for
Quantity
  • Availability
  • Add to Cart