ENA018106031 Enamine

5-amino-N-(4-chlorophenyl)-2-(morpholin-4-yl)benzene-1-sulfonamide

  • MDL number MFCD02706749
  • Cas Number : 327072-90-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 195-196
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-02364

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02364
Availability details for
Quantity
  • Availability
  • Add to Cart