ENA018106032 Enamine

3-(azepane-1-sulfonyl)aniline

  • MDL number MFCD03147234
  • Cas Number : 91619-39-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 75-76
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-02365

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02365
Availability details for
Quantity
  • Availability
  • Add to Cart