ENA018106037 Enamine

3-amino-N-(2,5-dichlorophenyl)-4-(diethylamino)benzene-1-sulfonamide

  • MDL number MFCD03147236
  • Cas Number : 379254-28-1
  • Molecular Weight : 388.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02370

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02370
Availability details for
Quantity
  • Availability
  • Add to Cart