ENA018106044 Enamine

3-Amino-N-(2-chloro-phenyl)-4-pyrrolidin-1-yl-benzenesulfonamide

  • MDL number MFCD02708586
  • Cas Number : 327106-71-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02379

Safety Information

msds https://www.enaminestore.com/catalog/EN300-02379
Availability details for
Quantity
  • Availability
  • Add to Cart