ENA018106052 Enamine

5-Amino-N-(2,5-dichloro-phenyl)-2-morpholin-4-yl-benzenesulfonamide

  • MDL number MFCD02709732
  • Cas Number : 329268-50-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 66...68
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02387

Safety Information

msds https://www.enaminestore.com/catalog/EN300-02387
Availability details for
Quantity
  • Availability
  • Add to Cart