ENA018106071 Enamine

4-methoxy-3-(piperidine-1-sulfonyl)aniline

  • MDL number MFCD03964197
  • Cas Number : 726157-16-0
  • Molecular Weight : 270.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-02406

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02406
Availability details for
Quantity
  • Availability
  • Add to Cart