ENA018106073 Enamine

2-methoxy-5-(piperidine-1-sulfonyl)aniline

  • MDL number MFCD03147336
  • Cas Number : 379255-14-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02408

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02408
Availability details for
Quantity
  • Availability
  • Add to Cart