ENA018106076 Enamine

2-(4-aminophenyl)-1H-1,3-benzodiazol-6-amine

  • MDL number MFCD00043996
  • Cas Number : 7621-86-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 198-199
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02411

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02411
Availability details for
Quantity
  • Availability
  • Add to Cart