ENA018106079 Enamine

4-methoxy-3-(morpholine-4-sulfonyl)aniline

  • MDL number MFCD03970369
  • Cas Number : 554407-02-2
  • Molecular Weight : 272.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-02415

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02415
Availability details for
Quantity
  • Availability
  • Add to Cart