ENA018106081 Enamine

3-[(4-fluorophenyl)amino]propanenitrile

  • MDL number MFCD01910095
  • Cas Number : 380190-11-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 47-48
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02417

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02417
Availability details for
Quantity
  • Availability
  • Add to Cart