ENA018106084 Enamine

3-[(2-methoxyphenyl)amino]propanenitrile

  • MDL number MFCD00460649
  • Cas Number : 27472-15-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-02420

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02420
Availability details for
Quantity
  • Availability
  • Add to Cart