ENA018106089 Enamine

N-(3-methoxyphenyl)-1,3-benzothiazol-2-amine

  • MDL number MFCD00115420
  • Cas Number : 216502-43-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 117-118
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02425

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02425
Availability details for
Quantity
  • Availability
  • Add to Cart