ENA018106106 Enamine

1-(1,3-benzoxazol-2-yl)-3-methyl-1H-pyrazol-5-amine

  • MDL number MFCD06380346
  • Cas Number : 926190-29-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-02449

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02449
Availability details for
Quantity
  • Availability
  • Add to Cart