ENA018106113 Enamine

5-(4-Chloro-benzylsulfanyl)-[1,3,4]thiadiazol-2-ylamine

  • MDL number MFCD00114062
  • Cas Number : 72836-33-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02456

Safety Information

msds https://www.enaminestore.com/catalog/EN300-02456
Availability details for
Quantity
  • Availability
  • Add to Cart