ENA018106115 Enamine

N-(2-methoxy-5-methylphenyl)-5,6-dihydro-4H-1,3-thiazin-2-amine

  • MDL number MFCD03150733
  • Cas Number : 91557-27-6
  • Molecular Weight : 236.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-02458

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02458
Availability details for
Quantity
  • Availability
  • Add to Cart