ENA018106126 Enamine

2-amino-N-[(furan-2-yl)methyl]benzamide

  • MDL number MFCD02374151
  • Cas Number : 117507-63-8
  • Molecular Weight : 216.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 78-79
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-02469

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02469
Availability details for
Quantity
  • Availability
  • Add to Cart