ENA018106140 Enamine

N-{4-[(difluoromethyl)sulfanyl]phenyl}-1,3-benzothiazol-2-amine

  • MDL number MFCD03150325
  • Cas Number : 380349-37-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-02483

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02483
Availability details for
Quantity
  • Availability
  • Add to Cart