ENA018106141 Enamine

4-ethyl-N-(4-ethylphenyl)-4,5-dihydro-1,3-thiazol-2-amine

  • MDL number MFCD03150323
  • Cas Number : 380349-33-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02484

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02484
Availability details for
Quantity
  • Availability
  • Add to Cart