ENA018106143 Enamine

N-[4-(5-methyl-1,3-benzothiazol-2-yl)phenyl]-5,6-dihydro-4H-1,3-thiazin-2-amine

  • MDL number MFCD03150682
  • Cas Number : 380431-15-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-02486

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02486
Availability details for
Quantity
  • Availability
  • Add to Cart