ENA018106146 Enamine

3,4-diamino-N-(2-methoxyphenyl)benzene-1-sulfonamide

  • MDL number MFCD03152344
  • Cas Number : 379725-41-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02489

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02489
Availability details for
Quantity
  • Availability
  • Add to Cart