ENA018106161 Enamine

N-(2-methoxy-5-methylphenyl)-4,4-dimethyl-4,5-dihydro-1,3-thiazol-2-amine

  • MDL number MFCD03152621
  • Cas Number : 379728-06-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-02505

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02505
Availability details for
Quantity
  • Availability
  • Add to Cart