ENA018106163 Enamine

N-(4-chlorophenyl)-1,3-benzothiazol-2-amine

  • MDL number MFCD03152613
  • Cas Number : 6276-78-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02507

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02507
Availability details for
Quantity
  • Availability
  • Add to Cart