ENA018106165 Enamine

N-(2-ethoxyphenyl)-1,3-benzothiazol-2-amine

  • MDL number MFCD03152812
  • Cas Number : 379729-89-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-02509

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02509
Availability details for
Quantity
  • Availability
  • Add to Cart