ENA018106170 Enamine

1,8-diazatricyclo[7.5.0.0,2,7]tetradeca-2(7),3,5,8-tetraen-5-amine

  • MDL number MFCD03152428
  • Cas Number : 379726-16-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 180-181
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02514

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02514
Availability details for
Quantity
  • Availability
  • Add to Cart