ENA018106193 Enamine

5-[(4-bromophenyl)methyl]-1,3-thiazol-2-amine

  • MDL number MFCD01910159
  • Cas Number : 327062-46-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 123-125
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-02544

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02544
Availability details for
Quantity
  • Availability
  • Add to Cart